1-Nitro-4-(2-propynyloxy)benzene 97%
Dodavatel: Apollo Scientific
OR937255-1G
OR937255-5G
APOSOR937255-1GEA
6420
CZK
APOSOR937255-1G
APOSOR937255-5G
1-Nitro-4-(2-propynyloxy)benzene 97%
1-Nitro-4-(2-propynyloxy)benzene
Vzorec:
C₉H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD01142872 CAS číslo: 17061-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...