Phenylglyoxal monohydrate 95%
Dodavatel: Apollo Scientific
Synonyms:
2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
OR936846-25G
OR936846-5G
APOSOR936846-25GEA
6390
CZK
APOSOR936846-25G
APOSOR936846-5G
Phenylglyoxal monohydrate 95%
Phenylglyoxal monohydrate
Vzorec:
C₈H₆O₂·H₂O MW: 152.15 g/mol (134.12 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149499 CAS číslo: 78146-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...