Tisk…

(4-(Benzyl(methyl)carbamoyl)phenyl)boronic acid 96%

Dodavatel: Apollo Scientific
Varování

OR9361-1G OR9361-5G
APOSOR9361-1GEA 5000 CZK
APOSOR9361-1G APOSOR9361-5G
(4-(Benzyl(methyl)carbamoyl)phenyl)boronic acid 96%
(4-(Benzyl(methyl)carbamoyl)phenyl)boronic acid
Vzorec: C₁₅H₁₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08235067
CAS číslo: 874219-49-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR