Tisk…

Ethyl 6-oxospiro[2.5]octane-1-carboxylate

Dodavatel: Apollo Scientific

OR935842-250MG OR935842-500MG
APOSOR935842-250MGEA 26290 CZK
APOSOR935842-250MG APOSOR935842-500MG
Ethyl 6-oxospiro[2.5]octane-1-carboxylate
Ethyl 6-oxospiro[2.5]octane-1-carboxylate
Vzorec: C₁₁H₁₆O₃
MW: 196,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28135615
CAS číslo: 1447942-87-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR