Tisk…
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Dodavatel: Apollo Scientific
OR933912-1G OR933912-250MG OR933912-500MG
APOSOR933912-1GEA 42320 CZK
APOSOR933912-1G APOSOR933912-250MG APOSOR933912-500MG
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Methyl 3-(aminomethyl)cyclobutane-1-carboxylate hydrochloride
Vzorec: C₇H₁₃NO₂·ClH
MW: 179,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28014565
CAS číslo: 1630907-39-9