Tisk…

2,6-Diazaspiro[3.4]octane dihydrochloride

Dodavatel: Apollo Scientific

OR933563-1G OR933563-5G
APOSOR933563-1GEA 16440 CZK
APOSOR933563-1G APOSOR933563-5G
2,6-Diazaspiro[3.4]octane dihydrochloride
2,6-Diazaspiro[3.4]octane dihydrochloride
Vzorec: C₆H₁₂N₂·2ClH
MW: 185,095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28166283
CAS číslo: 1630906-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR