Tisk…

1-Methyl-1H-indole-7-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR933307-1G OR933307-250MG
APOSOR933307-1GEA 2900 CZK
APOSOR933307-1G APOSOR933307-250MG
1-Methyl-1H-indole-7-carboxylic acid 95%
1-Methyl-1H-indole-7-carboxylic acid
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09879957
CAS číslo: 167479-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR