Tisk…

4-Hydroxy-3-methylphenylacetic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(4-Hydroxy-3-methylphenyl)acetic acid

OR933292-1G OR933292-5G
APOSOR933292-1GEA 2940 CZK
APOSOR933292-1G APOSOR933292-5G
4-Hydroxy-3-methylphenylacetic acid 95%
4-Hydroxy-3-methylphenylacetic acid
Vzorec: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656118
CAS číslo: 29121-56-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR