2-Ethyl-5-nitropyridine 95%
Dodavatel: Apollo Scientific
OR933251-1G
OR933251-250MG
APOSOR933251-1GEA
6520
CZK
APOSOR933251-1G
APOSOR933251-250MG
2-Ethyl-5-nitropyridine 95%
2-Ethyl-5-nitropyridine
Vzorec:
C₇H₈N₂O₂ MW: 152.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03617763 CAS číslo: 31557-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...