1-Methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR933189-1G
OR933189-250MG
APOSOR933189-1GEA
3770
CZK
APOSOR933189-1G
APOSOR933189-250MG
1-Methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde 95%
1-Methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde
Vzorec:
C₁₁H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06738236 CAS číslo: 67735-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...