3,4-Dihydro-2H-chromen-3-ylamine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms:
Chroman-3-amine hydrochloride
OR933153-1G
OR933153-250MG
APOSOR933153-1GEA
9860
CZK
APOSOR933153-1G
APOSOR933153-250MG
3,4-Dihydro-2H-chromen-3-ylamine hydrochloride 95%
3,4-Dihydro-2H-chromen-3-ylamine hydrochloride
Vzorec:
C₉H₁₁NO·ClH MW: 185,651 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01736331 CAS číslo: 18518-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...