Tisk…

tert-Butyl n-(2-cyanoethyl)-N-methylcarbamate 95%

Dodavatel: Apollo Scientific

OR933135-1G OR933135-5G
APOSOR933135-1GEA 6620 CZK
APOSOR933135-1G APOSOR933135-5G
tert-Butyl n-(2-cyanoethyl)-N-methylcarbamate 95%
tert-Butyl n-(2-cyanoethyl)-N-methylcarbamate
Vzorec: C₉H₁₆N₂O₂
MW: 184,236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909493
CAS číslo: 128304-84-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR