Sodium 2-(hydroxymethylamino)acetate 95%
Dodavatel: Apollo Scientific
OR933097-1G
OR933097-250MG
APOSOR933097-1GEA
4630
CZK
APOSOR933097-1G
APOSOR933097-250MG
Sodium 2-(hydroxymethylamino)acetate 95%
Sodium 2-(hydroxymethylamino)acetate
Vzorec:
C₃H₆NO₃·Na Storage Temperature: Refrigerator |
MDL Number:
MFCD04307769 CAS číslo: 70161-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...