Tisk…

7-Chloro-1,2,3,4-tetrahydroquinoline ≥98%

Dodavatel: Apollo Scientific

OR933042-1G OR933042-5G
APOSOR933042-1GEA 2150 CZK
APOSOR933042-1G APOSOR933042-5G
7-Chloro-1,2,3,4-tetrahydroquinoline ≥98%
7-Chloro-1,2,3,4-tetrahydroquinoline
Vzorec: C₉H₁₀ClN
MW: 167,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544268
CAS číslo: 90562-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR