tert-Butyl (4-(chlorosulphonyl)phenyl)carbamate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR933008-1G
OR933008-5G
APOSOR933008-1GEA
4370
CZK
APOSOR933008-1G
APOSOR933008-5G
tert-Butyl (4-(chlorosulphonyl)phenyl)carbamate
tert-Butyl (4-(chlorosulphonyl)phenyl)carbamate
Vzorec:
C₁₁H₁₄ClNO₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD10697736 CAS číslo: 269747-25-3 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...