Tisk…

(S)-1-Benzyl-3-methylpiperazine 95%

Dodavatel: Apollo Scientific

OR932987-1G OR932987-250MG
APOSOR932987-1GEA 22950 CZK
APOSOR932987-1G APOSOR932987-250MG
(S)-1-Benzyl-3-methylpiperazine 95%
(S)-1-Benzyl-3-methylpiperazine
Vzorec: C₁₂H₁₈N₂
MW: 190,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062535
CAS číslo: 132871-12-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR