3-Bromo-1H-pyrazolo[3,4-b]pyrazine 95%
Dodavatel: Apollo Scientific
OR932957-1G
OR932957-250MG
APOSOR932957-1GEA
22950
CZK
APOSOR932957-1G
APOSOR932957-250MG
3-Bromo-1H-pyrazolo[3,4-b]pyrazine 95%
3-Bromo-1H-pyrazolo[3,4-b]pyrazine
Vzorec:
C₅H₃BrN₄ MW: 199,008 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834831 CAS číslo: 81411-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...