Tisk…

4-tert-Butyl-N-methylaniline 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(tert-Butyl)-N-methylaniline

OR932750-1G OR932750-250MG
APOSOR932750-1GEA 8830 CZK
APOSOR932750-1G APOSOR932750-250MG
4-tert-Butyl-N-methylaniline 95%
4-tert-Butyl-N-methylaniline
Vzorec: C₁₁H₁₇N
MW: 163,259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111157
CAS číslo: 5279-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR