Tisk…

4-Methyloxazole

Dodavatel: Apollo Scientific

OR932677-5G
APOSOR932677-5GEA 1620 CZK
APOSOR932677-5G
4-Methyloxazole
4-Methyloxazole
Vzorec: C₄H₅NO
MW: 83,089 g/mol
Bod varu: 86…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110237
CAS číslo: 693-93-6
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR