Tisk…

4-Carbethoxy-3-methyl-2-cyclohexen-1-one 96%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-methyl-4-oxocyclohex-2-enecarboxylate

OR932355-100G OR932355-25G
APOSOR932355-100GEA 6910 CZK
APOSOR932355-100G APOSOR932355-25G
4-Carbethoxy-3-methyl-2-cyclohexen-1-one 96%
4-Carbethoxy-3-methyl-2-cyclohexen-1-one
Vzorec: C₁₀H₁₄O₃
MW: 182,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001582
CAS číslo: 487-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR