Tisk…

Ethyl 1-bromocyclopropanecarboxylate 95%

Dodavatel: Apollo Scientific

OR932351-1G OR932351-250MG
APOSOR932351-1GEA 900 CZK
APOSOR932351-1G APOSOR932351-250MG
Ethyl 1-bromocyclopropanecarboxylate 95%
Ethyl 1-bromocyclopropanecarboxylate
Vzorec: C₆H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD01314227
CAS číslo: 89544-83-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR