Boc-DL-tert-leucine 97%
Dodavatel: Apollo Scientific
OR932319-1G
OR932319-25G
OR932319-5G
APOSOR932319-1GEA
3480
CZK
APOSOR932319-1G
APOSOR932319-25G
APOSOR932319-5G
Boc-DL-tert-leucine 97%
Boc-DL-tert-leucine
Vzorec:
C₁₁H₂₁NO₄ MW: 231,289 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057782 CAS číslo: 169870-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...