(S)-2-Phenylpyrrolidine 95%
Dodavatel: Apollo Scientific
OR932248-1G
OR932248-250MG
APOSOR932248-1GEA
10880
CZK
APOSOR932248-1G
APOSOR932248-250MG
(S)-2-Phenylpyrrolidine 95%
(S)-2-Phenylpyrrolidine
Vzorec:
C₁₀H₁₃N MW: 147,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06762549 CAS číslo: 59347-91-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...