3-Aminomethyl-2-methoxypyridine hydrochloride 95%
Dodavatel: Apollo Scientific
OR932171-1G
OR932171-250MG
APOSOR932171-1GEA
12830
CZK
APOSOR932171-1G
APOSOR932171-250MG
3-Aminomethyl-2-methoxypyridine hydrochloride 95%
3-Aminomethyl-2-methoxypyridine hydrochloride
Vzorec:
C₇H₁₀N₂O·ClH MW: 174.628 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23703574 CAS číslo: 1588441-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...