Tisk…

2,3-Dimethyl-1H-indole-7-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR932160-1G OR932160-250MG
APOSOR932160-1GEA 20270 CZK
APOSOR932160-1G APOSOR932160-250MG
2,3-Dimethyl-1H-indole-7-carbaldehyde 95%
2,3-Dimethyl-1H-indole-7-carbaldehyde
Vzorec: C₁₁H₁₁NO
MW: 173,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059725
CAS číslo: 103987-28-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR