5-Chloro-3-ethyl-1-methyl-1H-pyrazole-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR932091-1G
OR932091-250MG
APOSOR932091-1GEA
11380
CZK
APOSOR932091-1G
APOSOR932091-250MG
5-Chloro-3-ethyl-1-methyl-1H-pyrazole-4-carbaldehyde 95%
5-Chloro-3-ethyl-1-methyl-1H-pyrazole-4-carbaldehyde
Vzorec:
C₇H₉ClN₂O MW: 172,612 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788964 CAS číslo: 128564-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...