Tisk…

(R)-S-tert-Butyl 2-methylpropane-2-sulphinothioate 95%

Dodavatel: Apollo Scientific

OR932024-1G OR932024-250MG
APOSOR932024-1GEA 5460 CZK
APOSOR932024-1G APOSOR932024-250MG
(R)-S-tert-Butyl 2-methylpropane-2-sulphinothioate 95%
(R)-S-tert-Butyl 2-methylpropane-2-sulphinothioate
Vzorec: C₈H₁₈OS₂
Storage Temperature: Ambient
MDL Number: MFCD08460193
CAS číslo: 67734-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR