Tisk…

(R)-3-Cyclobutylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: (R)-2-Amino-3-cyclobutylpropanoic acid

OR931954-1G OR931954-250MG
APOSOR931954-1GEA 9200 CZK
APOSOR931954-1G APOSOR931954-250MG
(R)-3-Cyclobutylalanine 97%
(R)-3-Cyclobutylalanine
Vzorec: C₇H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09264348
CAS číslo: 174266-00-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR