(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-acetic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)acetic acid
OR931801-1G
OR931801-5G
APOSOR931801-1GEA
2440
CZK
APOSOR931801-1G
APOSOR931801-5G
(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-acetic acid 95%
(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-acetic acid
Vzorec:
C₁₀H₁₀O₄ MW: 194,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066923 CAS číslo: 17253-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...