Tisk…

4-Chloro-5-methyl-2-(methylthio)pyrimidine 95%

Dodavatel: Apollo Scientific

OR931785-1G OR931785-250MG
APOSOR931785-1GEA 3770 CZK
APOSOR931785-1G APOSOR931785-250MG
4-Chloro-5-methyl-2-(methylthio)pyrimidine 95%
4-Chloro-5-methyl-2-(methylthio)pyrimidine
Vzorec: C₆H₇ClN₂S
MW: 174,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951961
CAS číslo: 61044-96-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR