4-Bromo-5-methylisoxazole 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR931756-1G
OR931756-250MG
OR931756-5G
APOSOR931756-1GEA
6050
CZK
APOSOR931756-1G
APOSOR931756-250MG
APOSOR931756-5G
4-Bromo-5-methylisoxazole 96%
4-Bromo-5-methylisoxazole
Vzorec:
C₄H₄BrNO MW: 161,985 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD13689016 CAS číslo: 7064-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...