1-Vinyl-1H-pyrazole-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR931741-1G
OR931741-250MG
APOSOR931741-1GEA
15310
CZK
APOSOR931741-1G
APOSOR931741-250MG
1-Vinyl-1H-pyrazole-4-carbaldehyde 95%
1-Vinyl-1H-pyrazole-4-carbaldehyde
Vzorec:
C₆H₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD12198430 CAS číslo: 864723-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...