Tisk…

3-Formyl-1H-indole-6-carbonitrile ≥98%

Dodavatel: Apollo Scientific

OR931695-5G
APOSOR931695-5GEA 3910 CZK
APOSOR931695-5G
3-Formyl-1H-indole-6-carbonitrile ≥98%
3-Formyl-1H-indole-6-carbonitrile
Vzorec: C₁₀H₆N₂O
MW: 170,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657155
CAS číslo: 83783-33-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR