Tisk…

2,5-Dichlorophenethyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(2,5-Dichlorophenyl)ethan-1-ol

OR931635-10G OR931635-5G
APOSOR931635-10GEA 1740 CZK
APOSOR931635-10G APOSOR931635-5G
2,5-Dichlorophenethyl alcohol 95%
2,5-Dichlorophenethyl alcohol
Vzorec: C₈H₈Cl₂O
MW: 191,055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038828
CAS číslo: 1875-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR