Tisk…

tert-Butyl but-3-yn-1-ylcarbamate 95%

Dodavatel: Apollo Scientific

OR931520-1G
APOSOR931520-1GEA 1700 CZK
APOSOR931520-1G
tert-Butyl but-3-yn-1-ylcarbamate 95%
tert-Butyl but-3-yn-1-ylcarbamate
Vzorec: C₉H₁₅NO₂
MW: 169,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698150
CAS číslo: 149990-27-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR