Tisk…
2-(Chloromethyl)phenylacetic acid 95%
Dodavatel: Apollo Scientific
Synonyms: 2-(2-(Chloromethyl)phenyl)acetic acid
OR931497-1G OR931497-5G
APOSOR931497-1GEA 1950 CZK
APOSOR931497-1G APOSOR931497-5G
2-(Chloromethyl)phenylacetic acid 95%
2-(Chloromethyl)phenylacetic acid
Vzorec: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200800
CAS číslo: 95335-46-9