2-Sec-butoxybenzaldehyde 95%
Dodavatel: Apollo Scientific
OR931491-1G
OR931491-250MG
APOSOR931491-1GEA
2650
CZK
APOSOR931491-1G
APOSOR931491-250MG
2-Sec-butoxybenzaldehyde 95%
2-Sec-butoxybenzaldehyde
Vzorec:
C₁₁H₁₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02629688 CAS číslo: 22921-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...