Tisk…

5-Aminomethyl-thiophene-2-carbonitrile 95%

Dodavatel: Apollo Scientific

OR931478-1G OR931478-250MG
APOSOR931478-1GEA 11810 CZK
APOSOR931478-1G APOSOR931478-250MG
5-Aminomethyl-thiophene-2-carbonitrile 95%
5-Aminomethyl-thiophene-2-carbonitrile
Vzorec: C₆H₆N₂S
MW: 138,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457570
CAS číslo: 227279-10-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR