Tisk…

3-Chloro-5-hydroxy-2-methoxypyridine 95%

Dodavatel: Apollo Scientific

OR931387-1G OR931387-250MG
APOSOR931387-1GEA 23580 CZK
APOSOR931387-1G APOSOR931387-250MG
3-Chloro-5-hydroxy-2-methoxypyridine 95%
3-Chloro-5-hydroxy-2-methoxypyridine
Vzorec: C₆H₆ClNO₂
MW: 159,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190369
CAS číslo: 1196157-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR