Tisk…

5-Chloro-3-methyl-4-nitro-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR931371-1G
APOSOR931371-1GEA 2570 CZK
APOSOR931371-1G
5-Chloro-3-methyl-4-nitro-1H-pyrazole 95%
5-Chloro-3-methyl-4-nitro-1H-pyrazole
Vzorec: C₄H₄ClN₃O₂
MW: 161,546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462202
CAS číslo: 6814-58-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR