Tisk…

5-Cyano-3-pyridinecarboxylic acid methyl ester 95%

Dodavatel: Apollo Scientific

OR931210-1G OR931210-250MG
APOSOR931210-1GEA 21920 CZK
APOSOR931210-1G APOSOR931210-250MG
5-Cyano-3-pyridinecarboxylic acid methyl ester 95%
5-Cyano-3-pyridinecarboxylic acid methyl ester
Vzorec: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367902
CAS číslo: 106726-82-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR