Ethyl 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR931184-1G
OR931184-250MG
APOSOR931184-1GEA
19840
CZK
APOSOR931184-1G
APOSOR931184-250MG
Ethyl 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylate 95%
Ethyl 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Vzorec:
C₁₀H₁₃NO₃ MW: 195,215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00820157 CAS číslo: 16108-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...