2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
Dodavatel: Apollo Scientific
OR931181-1G
APOSOR931181-1GEA
3740
CZK
APOSOR931181-1G
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride
Vzorec:
C₇H₆ClNO·ClH MW: 192,043 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09033889 CAS číslo: 61889-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...