6-Chloroquinazolin-2-amine 95%
Dodavatel: Apollo Scientific
OR931131-1G
OR931131-250MG
APOSOR931131-1GEA
23970
CZK
APOSOR931131-1G
APOSOR931131-250MG
6-Chloroquinazolin-2-amine 95%
6-Chloroquinazolin-2-amine
Vzorec:
C₈H₆ClN₃ MW: 179,606 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368133 CAS číslo: 20028-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...