Tisk…

1-(2-Chloroethyl)-4-methylpiperazine hydrochloride 95%

Dodavatel: Apollo Scientific

OR930991-5G
APOSOR930991-5GEA 4530 CZK
APOSOR930991-5G
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride 95%
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride
Vzorec: C₇H₁₅ClN₂·ClH
MW: 199,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031872
CAS číslo: 126055-32-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR