Tisk…

1-Cyclopropyl-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR930988-1G OR930988-250MG
APOSOR930988-1GEA 20270 CZK
APOSOR930988-1G APOSOR930988-250MG
1-Cyclopropyl-1H-pyrrole-2-carbaldehyde 95%
1-Cyclopropyl-1H-pyrrole-2-carbaldehyde
Vzorec: C₈H₉NO
Storage Temperature: Ambient
MDL Number: MFCD12181369
CAS číslo: 1039758-67-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR