3-(Nitromethyl)cyclopentanone 95%
Dodavatel: Apollo Scientific
OR930987-1G
OR930987-250MG
APOSOR930987-1GEA
25000
CZK
APOSOR930987-1G
APOSOR930987-250MG
3-(Nitromethyl)cyclopentanone 95%
3-(Nitromethyl)cyclopentanone
Vzorec:
C₆H₉NO₃ MW: 143,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847689 CAS číslo: 81266-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...