Tisk…

2-Chloro-5-furan-2-yl-pyrazine 95%

Dodavatel: Apollo Scientific

OR930897-1G OR930897-250MG
APOSOR930897-1GEA 15310 CZK
APOSOR930897-1G APOSOR930897-250MG
2-Chloro-5-furan-2-yl-pyrazine 95%
2-Chloro-5-furan-2-yl-pyrazine
Vzorec: C₈H₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09033268
CAS číslo: 82619-63-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR