Tisk…

4-[Ethyl(methyl)amino]benzaldehyde 95%

Dodavatel: Apollo Scientific

OR930677-1G OR930677-250MG
APOSOR930677-1GEA 11380 CZK
APOSOR930677-1G APOSOR930677-250MG
4-[Ethyl(methyl)amino]benzaldehyde 95%
4-[Ethyl(methyl)amino]benzaldehyde
Vzorec: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD09864610
CAS číslo: 64693-47-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR