Methyl 2-chloro-5-methylbenzoate 95%
Dodavatel: Apollo Scientific
OR930591-5G
APOSOR930591-5GEA
2860
CZK
APOSOR930591-5G
Methyl 2-chloro-5-methylbenzoate 95%
Methyl 2-chloro-5-methylbenzoate
Vzorec:
C₉H₉ClO₂ MW: 184,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12025214 CAS číslo: 16220-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...